Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Flumazenil

Catalog No. 5059910001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-599-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-599-10001 Supplier MilliporeSigma™ Supplier No. 5059910001
Add to Cart
Add to Cart

A competitive blocker of benzodiazepine activation of inhibitory GABAergic synpases (ID50 = 200μg/kg/i

A competitive blocker of benzodiazepine activation of inhibitory GABAergic synpases (ID50 = 200μg/kg/i.p). Binds to the benzodiazepine site of GABAA receptors. Enhances GABAA-receptor mediated currents and antagonizes the enhancing effects of benzodiapine agonist flurazepam. Exhibits fast association and dissociation from the benzodiazepine binding site. Permeates the blood brain barrier, however, the level of uptake may depend on efflux effects of p-glycogprotein transporter.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins GABAA
Molecular Formula C15H14FN3O3
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.