Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Flurbiprofen

Catalog No. 344079100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-407-9100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-407-9100MG Supplier MilliporeSigma™ Supplier No. 344079100MG

missing translation for 'validMainframeMsg'

A mixture of S(+) and R(-) enantiomers.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by titration
Target Proteins Cyclooxygenase
CAS 5104-49-4
Molecular Formula C15H13FO2
Formula Weight 244.3g/mol
Solubility DMSO
Form Solid
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.