Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Focal Adhesion Kinase Inhibitor III

Catalog No. 5040450001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-404-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-404-50001 Supplier MilliporeSigma™ Supplier No. 5040450001
Add to Cart
Add to Cart

A cell-permeable pyrazolobenzothiazinedioxide compound that targets and locks FAK in its inactive conformation, resulting in preincubation time-dependent inhibition in kinase assays and FAK autophosphorylation in human prostate cancer PC3M-luc cells (IC50

A cell-permeable pyrazolobenzothiazinedioxide compound that targets and locks FAK in its inactive conformation, resulting in preincubation time-dependent inhibition in kinase assays and FAK autophosphorylation in human prostate cancer PC3M-luc cells (IC50 = 7.1μM with 30 min inhibitor preincubation), while exhibiting little activity toward Akt1, Aurora B, HER2, MEK1, p38α, Pyk2, or Src even at concentrations as high as 10μM.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Target Proteins FAK
Molecular Formula C22H26N4O2S
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.