Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Formyl Peptide Receptor-Like 1 Agonist, WKYMVm

Catalog No. 3442165MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-421-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-421-65MG Supplier MilliporeSigma™ Supplier No. 3442165MG
Add to Cart
Add to Cart

The Formyl Peptide Receptor-Like 1 Agonist, WKYMVm controls the biological activity of Formyl Peptide Receptor-Like 1. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Trp-Lys-Tyr-Met-Val-D-Met-CONH2
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Formyl Peptide Receptor-Like 1
Molecular Formula C41H61N9O7S2
Formula Weight 855.4g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.