Learn More
MilliporeSigma™ Calbiochem™ Formyl Peptide Receptor-Like 1 Antagonist, WRW4
Shop All MilliporeSigma ProductsDescription
- Packaged Under Inert Gas
- Peptide Sequence: H-Trp-Arg-Trp-Trp-Trp-Trp-CONH2
Specifications
Specifications
| Color | White |
| Formulation | Supplied as a trifluoroacetate salt. |
| Description | 95% by HPLC |
| Target Proteins | Formyl Peptide Receptor-Like 1 (FPRL1) |
| Molecular Formula | C61H65N15O6 |
| Formula Weight | 1104.3g/mol |
| Solubility | DMSO |
| Content And Storage | Hygroscopic |
| Form | Lyophilized |
| Quantity | 5 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.