Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ FTY720

Catalog No. 34459710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-459-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-459-710MG Supplier MilliporeSigma™ Supplier No. 34459710MG
Add to Cart
Add to Cart

A cell-permeable aminopropanediol immunosuppressive agent that displays lymphocyte sequestration properties

A cell-permeable aminopropanediol immunosuppressive agent that displays lymphocyte sequestration properties. Reported to be phosphorylated by sphingosine kinase to FTY720-P, which has been shown to potently stimulate GTPγS binding activity in S1P-transfected CHO cells (EC50 = 210 pM, 4.9 nM, 4.3 nM, and 1nM for S1P1, S1P3, S1P4 and S1P5, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by TLC
CAS 162359-56-0
Molecular Formula C19H33NO2·HCl
Formula Weight 343.9g/mol
Solubility DMSO or EtOH
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.