Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Furin Substrate, Fluorogenic

Catalog No. 3449355MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-493-55MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-493-55MG Supplier MilliporeSigma™ Supplier No. 3449355MG
Add to Cart
Add to Cart

A fluorogenic substrate for furin, a mammalian homolog of the yeast Kex2 endoprotease (kcat

A fluorogenic substrate for furin, a mammalian homolog of the yeast Kex2 endoprotease (kcat/Km ∽ 2 x 104 s-1M-1).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Fluorogenic substrate for furin
Molecular Formula C37H57N13O9
Formula Weight 827.9g/mol
Solubility H2O or DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.