Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Gβγ Modulator II, Gallein

Catalog No. 3717085MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-170-85MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-170-85MG Supplier MilliporeSigma™ Supplier No. 3717085MG
Add to Cart
Add to Cart

A cell-permeable xanthene compound that binds to Gβγ with high affinity (K

A cell-permeable xanthene compound that binds to Gβγ with high affinity (Kd ∽400nM derived from a surface plasma resonance binding study using Gβ1γ2) and blocks Gβγ-dependent cellular activities. Reported to exhibit in vivo prophylactic efficacy against carrageenan-induced neutrophil recruitment and tissue swelling in a murine paw edema model via either intraperitoneal or oral administration.
TRUSTED_SUSTAINABILITY

Specifications

Color Burgundy
Description 90% by HPLC
CAS 2103-64-2
Molecular Formula C20H12O7·2H2O
Formula Weight 400.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.