Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Gabapentin

Catalog No. 5085060001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-850-60001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-850-60001 Supplier MilliporeSigma™ Supplier No. 5085060001
Add to Cart
Add to Cart

A water soluble GABA analog that interacts with voltage-gated calcium channels in cortical neurons.

Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins α2δ subunit of L-type voltage gated Ca2+ channels
Molecular Formula C9H17NO2
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.