Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Galanthamine, Hydrobromide

Catalog No. 34567020MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-567-020MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-567-020MG Supplier MilliporeSigma™ Supplier No. 34567020MG
Add to Cart
Add to Cart

A competitive and reversible inhibitor of acetylcholinesterase.

A competitive and reversible inhibitor of acetylcholinesterase. Antimyasthenic agent. Can partially reverse the effects of scopolamine-induced amnesia in rats. Reported to improve learning and short-term memory in animal models.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Acetylcholinesterase
CAS 69353-21-5
Molecular Formula C17H21NO3·HBr
Formula Weight 368.3g/mol
Solubility DMSO, EtOH or H2O
Content And Storage Protect from moisture
Form Solid
Quantity 20 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.