Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GCase Activator, NCGC00188758

Catalog No. 5316600001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-166-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-166-00001 Supplier MilliporeSigma™ Supplier No. 5316600001
Add to Cart
Add to Cart

A cell-permeable pyrazolopyrimidine-carboxamide compound that directly binds to glucocerebrosidase (GCase; Kd ∽ 9μM for NT-467 fluorescent labeled GCase) and enhances its activity and serves as a chaperone for its translocation to the lysosomal compartment in fibroblasts of patients with Gaucher disease (AC50

A cell-permeable pyrazolopyrimidine-carboxamide compound that directly binds to glucocerebrosidase (GCase; Kd ∽ 9μM for NT-467 fluorescent labeled GCase) and enhances its activity and serves as a chaperone for its translocation to the lysosomal compartment in fibroblasts of patients with Gaucher disease (AC50 = 5.2 and 6.5μM for wild type and mutant N370S, respectively). Also shown to be effective in increasing GCase activity in macrophages from Gaucher's disease patients as well as in iPSC-derived macrophages (10.7 and 3.2 fold, respectively). Normalizes reactive oxygen species production and improves chemotaxis in Gaucher disease macrophages. Shown to permeate the blood-brain barrier. Exhibits desirable pharmacokinetic properties with plasma t1/2 = 19.3 h and Cmax of 1.03μM (following a single injection of 50mg/kg in mice).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Orange
Description 98% by HPLC
Target Proteins Glucocerebrosidase (Gcase)
Molecular Formula C17H14N4O
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.