Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Gemcitabine, HCl

Catalog No. 5045940001 Shop All MilliporeSigma Products
Change view
Click to view available options
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-459-40001 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-459-40001 Supplier MilliporeSigma™ Supplier No. 5045940001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A bioavailable, moderately toxic, deoxyctytidine analog with anti-metabolite and anti-tumor properties

A bioavailable, moderately toxic, deoxyctytidine analog with anti-metabolite and anti-tumor properties. Shown to be effective against a variety of tumors and blocks the growth of cancer cells in culture (IC 50 = 8.7, 57, 3.7, and 36nM for L1210, CEM, Caco2, and HeLa cells, respectively). Following its uptake into cells by nucleoside transporters it is phosphorylated to its mono (dFdCMP), di (dFdCDP), and triphosphorylated (dFdCTP) forms by deoxycytidine kinase. dFdCDP and dFdCTP are reported to inhibit the activity of ribonucleotide reductase and impede DNA synthesis and repair mechanisms and induce cell death. Blocks the progression of cells through the G1/S phase. The dFdCTP is shown to compete with endogenous deoxycytidine triphosphate (dCTP) for incorporation into DNA and against CTP into RNA. Gemcitabine is also shown to block mitochondrial DNA polymerase γ. Works synergistically with other chemotherapeutic agents to enhance their cytotoxicity. Its half life is reported to be 42 to 94 min (depending upon the age and sex of the individual).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins DNA
Molecular Formula C9H11F2N3O4·HCl
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.