Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GGACK

Catalog No. 3474365MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-743-65MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-743-65MG Supplier MilliporeSigma™ Supplier No. 3474365MG
Add to Cart
Add to Cart

An irreversible inhibitor of urokinase (IC50 1 μM) and Factor Xa.

Peptide Sequence: H-Glu-Gly-Arg-CMK
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 80% by HPLC
Target Proteins urokinase
Molecular Formula C14H25ClN6O5
Formula Weight 392.8g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from Moisture
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.