Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glucagon-Like Peptide 1 Receptor Agonist II

Catalog No. 34600710MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-600-710MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-600-710MG Supplier MilliporeSigma™ Supplier No. 34600710MG
Add to Cart
Add to Cart

A pyrimidine small molecule that acts as a GLP-1 allosteric agonist that induces cAMP signaling in HEK293 cells expressing the GLP-1 receptor with EC50

A pyrimidine small molecule that acts as a GLP-1 allosteric agonist that induces cAMP signaling in HEK293 cells expressing the GLP-1 receptor with EC50 = 0.66μM and 99% efficacy (similar data with rat GLP-1 receptor), does not compete with radiolabelled GLP-1 in receptor binding assays, and is not active in cells expressing the GLP-2, GIP, PTH, or glucagon receptors. In rodent islets, insulin secretion increases in a dose-dependent and glucose-dependent manner, either alone or in an additive fashion when combined with the endogenous GLP-1 peptide. In vivo , this molecule exhibits insulinotropic effects when dosed at 10mg/kg in SD rats undergoing an IVGTT and a hyperglycemic clamp. It is also shown to increase insulin secretion in perifusion assays involving normal and diabetic human islets ex vivo , at concentrations between 1 and 10μM.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description >95% by HPLC
Molecular Formula C20H17F3N2O2S
Formula Weight 406.42g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.