Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glutaminase Inhibitor, Compound 968

Catalog No. 35201010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-201-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 35-201-010MG Supplier MilliporeSigma™ Supplier No. 35201010MG
Add to Cart
Add to Cart

Glutaminase Inhibitor, Compd 968, is a cell-permeable, reversible inhibitor of mitochondrial glutaminase. Represses growth & invasive activity in glutaminase upregulated fibroblasts and tumor cells.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Molecular Formula C27H27BrN2O
Formula Weight 475.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.