Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glutaminase Inhibitor II, BPTES

Catalog No. 5300300001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-003-00001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-003-00001 Supplier MilliporeSigma™ Supplier No. 5300300001
Add to Cart
Add to Cart

A cell-permeable, potent, selective, reversible, and uncompetitive allosteric inhibitor of kidney-type glutaminase (GLS1). Does not affect the activity of liver glutaminase.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 94% by HPLC
Target Proteins kidney-type glutaminase
CAS 314045-39-1
Molecular Formula C24H24N6O2S3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.