Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glycogen, Molecular Biology Grade, from Mussel

Catalog No. 3615071ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-150-71ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-150-71ML Supplier MilliporeSigma™ Supplier No. 3615071ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable as carrier substance for precipitation of nucleic acids

  • OneμL is sufficient to precipitate picogram quantities of nucleic acid with Ethanol
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mL
Source Mussel
Product Type Glycogen, Molecular Biology Grade
Content And Storage -20°C
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.