Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Glycoprotein Deglycosylation Kit

Catalog No. 3622801KIT Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-228-01KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-228-01KIT Supplier MilliporeSigma™ Supplier No. 3622801KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Removes N-linked, simple complex O-linked oligosaccharides from glycoproteins in single reaction.

EMD Millipore Calbiochem™ Glycoprotein Deglycosylation Kit is suitable to use with native or denaturation protocols.

  • Neutral pH
  • No degradation of protein
  • Contains sequencing grade enzymes without azide, BSA, or glycerol
  • Can be used to deglycosylate up to 2mg glycoprotein
  • Packaging: Fibre Case
TRUSTED_SUSTAINABILITY

Specifications

Includes Bovine Fetuin Control, Denaturation Solution, Endo-α-N-Acetylgalactosaminidase, β1,4-Galactosidase, Glucosaminidase, N-Glycosidase F, Neuraminidase, Reaction Buffer, TRITON X-100 Detergent, and user protocol
Quantity 1 Kit
Total Assay Time 3 hr, to 5 days
Content And Storage 2°C to 8°C
Product Type Glycoprotein Deglycosylation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.