Learn More
Description
- Inhibits replication, adsorption, and penetration of severe acute respiratory syndrome-associated coronavirus (SARS-CV)
- Also known as 11β-hydroxysteroid dehydrogenase type 2 (11-HSD2) inhibitor
- Blocks growth of other viruses, including herpes virus
- Helps restore liver functions in patients with hepatitis C
Specifications
Specifications
| Content And Storage | -20°C |
| Form | Light yellow solid |
| Product Type | Antiviral |
| Quantity | 5 g |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.