Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Golgicide A

Catalog No. 34586210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-586-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-586-210MG Supplier MilliporeSigma™ Supplier No. 34586210MG
Add to Cart
Add to Cart

A cell-permeable quinoline compound that selectively targets ArfGEF GBF1, but not BIG1/2, and attenuates GBF1-mediated cellular vesicle traffickings in a reversible manner.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
CAS 1139889-93-2
Molecular Formula C17H14F2N2
Formula Weight 284.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.