Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
A phenylacetamide compound that acts as an allosteric agonist of FFA2 (GPR43), demonstrating a left-shifted acetate dose response (IC50 = 0.7μM) and 111% efficacy relative to acetate in hFFA2 forskolin-induced cAMP assays.
Spécifications
Spécifications
| Color | White |
| Description | >95% by HPLC |
| Molecular Formula | C21H21ClN2OS |
| Formula Weight | 384.92g/mol |
| Solubility | DMSO |
| Form | Solid |
| Quantity | 10 mg |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'