Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GR 127935 Hydrochloride

Catalog No. 5080140001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-801-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-801-40001 Supplier MilliporeSigma™ Supplier No. 5080140001
Add to Cart
Add to Cart

A selective 5-HT1B/1D receptor antagon

A selective 5-HT1B/1D receptor antagonist. Selectivity was found to be similar for B (pki = 9.2-9.8 )and D (pki = 8.6-9.1) receptor types. GR 127935 works well in vivo and was shown to block (+)-MDMA-induced hyperactivity in rats. It alters release of serotonin in the brain, as well as reducing drug-seeking behaviour in cocaine addicted rats. In transformed rat C5 glioma cells, this inhibitor blocked 5HT with an IC50 = 97 pM.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins 5-HT1B/1D
Molecular Formula C29H31N5O3·HCl
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.