Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Granzyme B Inhibitor IV

Catalog No. 3680561MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-805-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-805-61MG Supplier MilliporeSigma™ Supplier No. 3680561MG
Add to Cart
Add to Cart

The Granzyme B Inhibitor IV controls the biological activity of Granzyme B. This small molecule/inhibitor is primarily used for Cancer applications.

Peptide Sequence: Ac-Ile-Glu-Pro-Asp-CHO
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins granzyme B
Molecular Formula C22H34N4O9
Formula Weight 498.5g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.