Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Guanabenz Acetate

Catalog No. 370625100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-062-5100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-062-5100MG Supplier MilliporeSigma™ Supplier No. 370625100MG

A cell-permeable α2-adrenoceptor agonist that lowers protein misfolding in stressed cells and acts as a proteostatis regulator.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
CAS 23256-50-0
Molecular Formula C8H8Cl2N4·C2H4O2
Formula Weight 291.1g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.