Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Guanosine 3',5'-cyclic Monophosphate, N2,2'-O-Dibutyryl-, Sodium Salt

Catalog No. 37066010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-066-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-066-010MG Supplier MilliporeSigma™ Supplier No. 37066010MG
Add to Cart
Add to Cart

Cell-permeable cGMP analog that preferentially activates protein kinase G.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins Cell-permeable cGMP analog that preferentially activates protein kinase G.
CAS 51116-00-8
Molecular Formula C18H23N5O9P·Na
Formula Weight 507.4g/mol
Solubility EtOH or H2O
Content And Storage Hygroscopic
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.