Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Guanosine 3',5'-cyclic Monophosphate, Sodium Salt

Catalog No. 37065650MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-065-650MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-065-650MG Supplier MilliporeSigma™ Supplier No. 37065650MG
Add to Cart
Add to Cart

Important intracellular second messenger implicated in the action of nitric oxide in many cell types.

Important intracellular second messenger implicated in the action of nitric oxide in many cell types. Activates protein kinase G. Suppresses apoptosis in rat ovarian follicles.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description >98% by HPLC
Target Proteins Activates protein kinase G
CAS 40732-48-7
Molecular Formula C10H11N5O7P·Na
Formula Weight 367.2g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.