Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Guanosine 3',5'-cyclic Monophosphorothioate, 8-(4-Chlorophenylthio)-, Rp-Isomer, Triethylammonium Salt

Catalog No. 3706771UMOL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 μmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-067-71UMOL 1 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-067-71UMOL Supplier MilliporeSigma™ Supplier No. 3706771UMOL
Add to Cart
Add to Cart

A potent, cell-permeable, and metabolically-stable inhibitor of protein kinase G Iα, Iβ, and type II.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 99% by HPLC
Target Proteins PKG1α
Molecular Formula C16H15ClN5O6PS2·C6H15N
Formula Weight 605.1g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 μmol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.