Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Guanosine 3',5'-cyclic Monophosphorothioate, β-Phenyl-1,N2-etheno-8-bromo-, Rp-Isomer, Sodium Salt

Catalog No. 3706791UMOL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 μmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-067-91UMOL 1 μmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-067-91UMOL Supplier MilliporeSigma™ Supplier No. 3706791UMOL
Add to Cart
Add to Cart

A metabolically-stable, competitive inhibitor of protein kinase G types 1α and 1β (Ki = 30 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins PKG1α & 1β
CAS 172806-20-1
Molecular Formula C18H14BrN5O6PS·Na
Formula Weight 562.3g/mol
Solubility H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 μmol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.