Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ H-89, Dihydrochloride

Catalog No. 3719631MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-196-31MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-196-31MG Supplier MilliporeSigma™ Supplier No. 3719631MG
Add to Cart
Add to Cart

A cell-permeable, selective, reversible, ATP-competitive and potent inhibitor of protein kinase A (Ki = 48 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Clear to Off-white
Description 99% by HPLC
Target Proteins PKA
CAS 127243-85-0
Molecular Formula C20H20BrN3O2S·2HCl
Formula Weight 519.3g/mol
Solubility DMSO or H2O
Form Crystalline Film
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.