Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Haspin Kinase Inhibitor, CHR-6494

Catalog No. 37204010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-204-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-204-010MG Supplier MilliporeSigma™ Supplier No. 37204010MG
Add to Cart
Add to Cart

A cell-permeable indazolylimidazopyridazinamine compound that acts as a potent and reversible inhibitor of histone kinase haspin activity (IC50 = 2nM) with moderate selectivity over TrkA, GSK-3β, PIM1, Cdk1/B and Cdk2/A (% inhibition at 100nM = 58, 48, 36, 34 and 33, respectively) among a panel of 27-kina

A cell-permeable indazolylimidazopyridazinamine compound that acts as a potent and reversible inhibitor of histone kinase haspin activity (IC50 = 2nM) with moderate selectivity over TrkA, GSK-3β, PIM1, Cdk1/B and Cdk2/A (% inhibition at 100nM = 58, 48, 36, 34 and 33, respectively) among a panel of 27-kinases. Shown to block pH3-Thr3, with no effect on pH3-Ser10 and pH3-Ser28, cause mitotic catastrophe, upregulate BUB1 and cyclin B1 levels, and potently induce apoptosis (IC50 = 473, 500 and 752nM in HeLa, HCT-116 and MDA-MB-231 cells, respectively). Further, efficiently blocks bFGF-induced sprouting vessel by 70% at 1μM (chicken embryo aortic arch ring assay) and suppresses tumor growth in HCT-116 xenografted mouse model (50mg/kg, i.p.).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
CAS 1333377-65-3
Molecular Formula C16H16N6
Formula Weight 292.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.