Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- A complete assay system designed to measure histone deacetylase (HDAC) activity in cell or nuclear extracts, immunoprecipitates, or purified enzymes
- It has been used successfully with preparations of all class I HDACs-HDAC1, HDAC2, HDAC3 and HDAC8 - with class II HDACs 4-7, 9 and 10 and with yeast Sir2 and its human homolog, SIRT1
Specifications
Specifications
| Content And Storage | Store at -70°C; Store both of the 1/2 Volume Microplates at room temperature |
| Format | 96-well Plate |
| Sample Type | Cell Extracts, Immunoprecipitates, Purified Enzymes |
| For Use With (Application) | To measure histone deacetylase (HDAC) activity in cell or nuclear extracts, immunoprecipitates, or purified enzymes |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?