Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Heat Shock Protein Inhibitor I

Catalog No. 3732605MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-326-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-326-05MG Supplier MilliporeSigma™ Supplier No. 3732605MG
Add to Cart
Add to Cart

Heat Shock Protein Inhibitor I, CAS 218924-25-5 blocks the induction of HSP70, HSP72, and HSP105. Blocks the development of thermotolerance in a dose-dependent manner.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White to Light Yellow
Description 95% by HPLC
Target Proteins Hsp70
CAS 218924-25-5
Molecular Formula C13H11NO4
Formula Weight 245.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.