Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Hec1/Nek2 Mitotic Pathway Inhibitor I, INH1

Catalog No. 37327010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-327-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-327-010MG Supplier MilliporeSigma™ Supplier No. 37327010MG
Add to Cart
Add to Cart

The Hec1/Nek2 Mitotic Pathway Inhibitor I, INH1, also referenced under CAS 313553-47-8, controls the biological activity of Hec1/Nek2 Mitotic Pathway.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
CAS 313553-47-8
Molecular Formula C18H16N2OS
Formula Weight 308.4g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.