Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ HIF Inhibitor V

Catalog No. 5004980001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-049-80001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-049-80001 Supplier MilliporeSigma™ Supplier No. 5004980001
Add to Cart
Add to Cart

A cell-permeable phenoxyacrylamido compound that inhibits hypoxia-induced cellular HIF-1α accumulation in a manner similar to LW6 (Cat

A cell-permeable phenoxyacrylamido compound that inhibits hypoxia-induced cellular HIF-1α accumulation in a manner similar to LW6 (Cat. No. 400083) by upregulating Hippel-Lindau tumor suppressor gene product pVHL, but with much higher potency (IC50 = 120nM vs 2.44μM, respectively, against hypoxia-induced HRE reporter activity in HCT116 cells). Shown to suppress HIF-1 target genes expression in HCT116 cultures and inhibit HUVEC tubule formation (effective conc. ≥2μM) under hypoxia conditions.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Molecular Formula C32H38N2O5
Formula Weight 530.7g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.