Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ HIF Inhibitor VII

Catalog No. 5043790001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-437-90001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-437-90001 Supplier MilliporeSigma™ Supplier No. 5043790001
Add to Cart
Add to Cart

A cell-permeable benzoxadiazolamine that prevents HIF-2α-HIF-β/ARNT heterodimeration by perturbing HIF-2α Per-ARNT-Sim/PAS-B domain surface β-sheet conformation via a 1:1 stoichiometric binding ( K D = 81nM), while exhibiting no affinity toward HIF-1αPAS-B

A cell-permeable benzoxadiazolamine that prevents HIF-2α-HIF-β/ARNT heterodimeration by perturbing HIF-2α Per-ARNT-Sim/PAS-B domain surface β-sheet conformation via a 1:1 stoichiometric binding ( K D = 81nM), while exhibiting no affinity toward HIF-1αPAS-B domain. Shown to be metabolically stable and non-cytotoxic in cultures and selectively inhibit hypoxia-induced HIF-2α DNA binding and HIF-2α-dependent EPO mRNA upregulation (1 to10μM for 6 to 12 h), but not HIF-1α DNA binding or HIF-1α-dependent PGK1 mRNA upregulation, in Hep3B cells. Unlike HIF Inhibitors I-VI, this compound does not affect the cellular protein or mRNA levels of HIF-1α & HIF-2α.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 99% by HPLC
Target Proteins HIF-2α
Molecular Formula C12H6ClFN4O3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.