Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ HIF Prolyl Hydroxylase Inhibitor

Catalog No. 40008410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-008-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-008-410MG Supplier MilliporeSigma™ Supplier No. 40008410MG
Add to Cart
Add to Cart

A cell-permeable, hydroxyquinoline HIF Prolyl Hydroxylase (PHD) inhibitor that displays an IC50 = 2μM in a cell-based assay using a reporter produced by fusing HIF-1α oxygen degradable domain (ODD) to lucifer

A cell-permeable, hydroxyquinoline HIF Prolyl Hydroxylase (PHD) inhibitor that displays an IC50 = 2μM in a cell-based assay using a reporter produced by fusing HIF-1α oxygen degradable domain (ODD) to luciferase. This compound (5μM) is shown to significantly stabilize HIF-1α in vitro and upregulate known HIF target genes, Epo and VEGF, in SH-SY5Y human neuroblastoma cell lines. It also exerts a neuroprotective effect (IC50 = 0.25μM in an oxidative stress model using cortical neurons.
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 95% by HPLC
CAS 385786-48-1
Molecular Formula C21H16ClN3O
Formula Weight 377.82g/mol
Solubility DMSO
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.