Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Histone Acetyltransferase Inhibitor IV, CPTH2

Catalog No. 38211110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-211-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-211-110MG Supplier MilliporeSigma™ Supplier No. 38211110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable thiazolyl-hydrazone compound that inhibits the acetylation of histone H3 by recombinant human Gcn5 in a substrate-competive manner, while exhibiting little activity against rhGcn5 autoacetylation

A cell-permeable thiazolyl-hydrazone compound that inhibits the acetylation of histone H3 by recombinant human Gcn5 in a substrate-competive manner, while exhibiting little activity against rhGcn5 autoacetylation. Although functional knockout of Gcn5 in Saccharomyces cerevisiae by genetic deletion or by CPTH2 treatment is not detrimental to the survival of the budding yeast, CPTH2 treatment is shown to inhibit the growth of yeast strain genetically deleted of Gcn5, but not strains deleted of Elp3 or Sas2.
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description 95% by HPLC
CAS 357649-93-5
Molecular Formula C14H14ClN3S
Formula Weight 291.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.