Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Histone Acetyltransferase Inhibitor VIII, NU9056

Catalog No. 5005110001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-051-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-051-10001 Supplier MilliporeSigma™ Supplier No. 5005110001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable disulfane compound that is reported to be more potent against KAT5/Tip60 than PCAF, p300, and GCN5 in cell-free HAT activity assays (IC50 ≤2, ≤36, ≤60, and >100μM, respectively) and effectively reduce H3K14, H4K8, H4K16, but not α-tubulin, acetylation level in LNCaP prostate cancer cells (max inhibition at 5-10

A cell-permeable disulfane compound that is reported to be more potent against KAT5/Tip60 than PCAF, p300, and GCN5 in cell-free HAT activity assays (IC50 ≤2, ≤36, ≤60, and >100μM, respectively) and effectively reduce H3K14, H4K8, H4K16, but not α-tubulin, acetylation level in LNCaP prostate cancer cells (max inhibition at 5-10μM). Shown to induce cell cycle arrest and apoptosis (17 to 24μM) in LNCaP cultures and exhibit antiproliferation activity in both androgen-responsive and androgen-independent CaP cultures (GI50 from 7.5 to 27μM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish Brown
Description 97% by HPLC
Target Proteins KAT5
Molecular Formula C6H4N2S4
Solubility DMSO
Content And Storage Protect from light
Form Oil
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.