Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Histone Acetyltransferase p300 Inhibitor, C646

Catalog No. 38211310MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-211-310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-211-310MG Supplier MilliporeSigma™ Supplier No. 38211310MG
Add to Cart
Add to Cart

Histone Acetyltransferase p300 Inhibitor, C646, CAS 328968-36-1, is a cell-permeable, reversible inhibitor of p300/CBP HAT (Ki = 400nM). Competes with acetyl-CoA for the p300 Lys-CoA binding pocket.

A reversible, cell-permeable pyrazolone p300/CBP HAT inhibitor (Ki = 400nM), which competes with acetyl-CoA for the p300 Lys-CoA binding pocket. This compound demonstrates 86% inhibition against p300 at 10 nM, but elicits less than 10% inhibition against serotonin N-acetyltransferase, PCAF histone acetyltransferase, GCN5 histone acetyletransferase, Rtt109 histone acetyletransferase, Sas histone acetyl transferase, and MOZ histone acetyltransferase in a chemical screening assay. Treatment of C3H 10T1/2 mouse fibroblasts with 25μM of compound results in an inhibitory effect against basal and TSA-inducible acetylation of histones H3 and H4. It is also shown to inhibit human cell growth in both melanoma and non-small-cell-lung (NSCL) cancer cell lines at 10μM with similar or higher potency compared with that of the peptide-based bisubstrate p300/CBP HAT inhibitor Lys-CoA-Tat at 25μM.
TRUSTED_SUSTAINABILITY

Specifications

Color Brick Red
Description 99% by HPLC (sum of isomers)
Molecular Formula C24H19N3O6
Formula Weight 445.42g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.