Learn More
MilliporeSigma™ Calbiochem™ Histone Deacetylase Inhibitor III
Catalog No. 3821491MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
38-214-91MG 1 mg
1 options
Catalog No. 38-214-91MG Supplier MilliporeSigma™ Supplier No. 3821491MG
Add to Cart
Add to Cart
The Histone Deacetylase Inhibitor III, also referenced under CAS 251456-60-7, controls the biological activity of Histone Deacetylase. This small molecule/inhibitor is primarily used for Cell Structure applications.
A cell-permeable amide analog of Trichostatin A (Cat. No. 647925) that potently inhibits histone deacetylases (IC50 = 40nM for rat liver HDAC and IC50 = 100nM for maize HD). Induces differentiation and inhibits proliferation (∽2μM) of murine erythroleukemia cells.
TRUSTED_SUSTAINABILITY
Specifications
CAS 251456-60-7
Quantity 1 mg