Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Histone Deacetylase Inhibitor VI, HNHA

Catalog No. 38217210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-217-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-217-210MG Supplier MilliporeSigma™ Supplier No. 38217210MG
Add to Cart
Add to Cart

The Histone Deacetylase Inhibitor VI, HNHA, also referenced under CAS 926908-04-5, controls the biological activity of Histone Deacetylase.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
CAS 926908-04-5
Molecular Formula C17H21NO2S
Formula Weight 303.4g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.