Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Histone Lysine Demethylase Inhibitor VIII, GSK-J4

Catalog No. 42020510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-020-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-020-510MG Supplier MilliporeSigma™ Supplier No. 42020510MG
Add to Cart
Add to Cart

Histone Lysine Demethylase Inhibitor VIII, GSK-J4, CAS 1373423-53-0, is converted to GSK-J1 intracellularly for effective JMJD3 and UTX inhibition (∽ 25 to 50μM in HeLa cultures).

A cell-permeable GSK-J1 (Cat. No. 420204) prodrug that is ineffective in cell-free assays, but is effectively converted to GSK-J1 intracellularly for effective JMJD3 and UTX inhibition (effective conc. 25 to 50μM in HeLa cultures). GSK-J4 treatment is shown to block LPS-induced TNF-α production in primary human macrophage cultures (IC50 = 9μM by ELISA).

This probe is supplied in conjunction with the Structural Genomics Consortium (SGC). For further characterization details, please visit the SGC website (http://www.thesgc.org/scientists/chemical_probes/GSKJ1).
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 98% by HPLC
Target Proteins H3K27me3 demethylases JMJD3 & UTX
Molecular Formula C24H27N5O2
Formula Weight 417.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Semi-solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.