Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Histone Lysine Methyltransferase Inhibitor

Catalog No. 3821905MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-219-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-219-05MG Supplier MilliporeSigma™ Supplier No. 3821905MG
Add to Cart
Add to Cart

The Histone Lysine Methyltransferase Inhibitor, also referenced under CAS 935693-62-2, controls the biological activity of Histone Lysine Methyltransferase. This small molecule/inhibitor is primarily used for Cancer applications.

A cell-permeable diazepinyl-quinazolinamine, non-SAM (S-adenosylmethionine) analog-based HMTase (histone methyltransferase) inhibitor that selectively interferes with the G9a-catalyzed H3K9me2 (histone H3 Lys9 dimethylation) modification (IC50 = 1.7μM) in a reversible manner. It inhibits the GLP-catalyzed H3K9me3 only at much higher concentrations (IC50 = 38μM) and exhibits little activity against H3 methylations catalyzed by other HMTases (PRMT1, SET7/9, ESET, SUV39H1). Shown to effectively synergize with Oct 3/4 and Klf 4 in inducing reprogramming of primary murine fetal NPCs (Neural Progenitor Cells) into iPS (induced Pluripotent Stem) cells without additional viral transduction of Sox2 and c- Myc .
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
CAS 935693-62-2
Molecular Formula C28H38N6O2
Formula Weight 490.6g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.