Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ HMTase Inhibitor IX, MM-102

Catalog No. 5006490001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-064-90001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-064-90001 Supplier MilliporeSigma™ Supplier No. 5006490001
Add to Cart
Add to Cart

A cell-permeable peptidomimetic that prevents MLL1 (Mixed Lineage Leukemia 1) from complex assembling with WDR5 (Trp-Asp Repeat Domain 5), RbBP5 (Retinoblastoma Binding Protein 5), and ASH2L (Absent Small or Homeotic-2-Like) for enhanced H3K4 methyltransferase activity (IC50

A cell-permeable peptidomimetic that prevents MLL1 (Mixed Lineage Leukemia 1) from complex assembling with WDR5 (Trp-Asp Repeat Domain 5), RbBP5 (Retinoblastoma Binding Protein 5), and ASH2L (Absent Small or Homeotic-2-Like) for enhanced H3K4 methyltransferase activity (IC50 = 0.4nM). Shown to reduce HoxA9 and Meis-1 mRNA expression in myeloblasts derived from MLL1-AF9 -transduced murine bone marrow cells (50μM for 96 h) and preferentially inhibit the growth of MLL1-AF4-harboring MV4;11 and MLL1-ENL-harboring KOPN8 leukemia cultures (GI50 in 7 days = 25μM) via apoptosis induction, while exhibiting much reduced potency against BCR-ABL-dependent K562 cells (GI50 in 7 days = 84μM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
Description 95% by HPLC
Target Proteins WDR5
Molecular Formula C35H49F2N7O4·CF3CO2H
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.