Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ HNMPA-(AM)3

Catalog No. 3971005MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-710-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 39-710-05MG Supplier MilliporeSigma™ Supplier No. 3971005MG
Add to Cart
Add to Cart

A cell-permeable analog of HNMPA that yields the parent compound upon cleavage by cytosolic esterases.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Description 98% by TLC
Target Proteins tyrosine kinase
CAS 120944-03-8
Molecular Formula C20H23O10P
Formula Weight 454.4g/mol
Solubility DMSO or EtOH
Form Oil
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.