Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Hsp25 Kinase Inhibitor

Catalog No. 3858801MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-588-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-588-01MG Supplier MilliporeSigma™ Supplier No. 3858801MG
Add to Cart
Add to Cart

The Hsp25 Kinase Inhibitor controls the biological activity of Hsp25 KInase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Peptide Sequence: H-Lys-Lys-Lys-Ala-Leu-Asn-Arg-Gln-Leu-Gly-Val-Ala-Ala-OH
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 98% by HPLC
Target Proteins (Hsp25) kinase
Molecular Formula C61H113N21O16
Formula Weight 1396.7g/mol
Solubility DMSO or H2O
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.