Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Hsp70 Activator, YM-1

Catalog No. 5006150001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-061-50001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-061-50001 Supplier MilliporeSigma™ Supplier No. 5006150001
Add to Cart
Add to Cart

A stable and soluble MKT-077 analog that acts an allosteric activator of heat-shock protein 70 (HSP70)

A stable and soluble MKT-077 analog that acts an allosteric activator of heat-shock protein 70 (HSP70). Its action appears to be similar to HSP70 interacting protein (HIP). Shown to convert HSP70 to its tight-affinity conformation and promote the accumulation of the ADP-bound form of HSP70. Increases the binding of HSP70 to its unfolded substrates . Also shown to enhance ubiquitination and degradation of nNOS and polyglutamine androgen receptor (polyQ AR) (at ∽1 mM). Displays selective cytotoxicity across multiple cancer cell lines without affecting the normal cells. Also reported to restore tamoxifen sensitivity to a refractory tamoxifen-resistant MCF7 cells (∽10 mM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color Orange-red
Description 98% by HPLC
Target Proteins HSP70
Molecular Formula C20H19N3OS2·HCl
Formula Weight 417.98g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.