Learn More
Description
- Member of β-galactoside-binding animal lectins
- Levels of galectin-3 have been reported to be elevated in several metastatic cancers
Specifications
Specifications
| Formulation | Lyophilized from distilled water |
| Quantity | 100 μg |
| Storage Requirements | -20°C |
| Endotoxin Concentration | ≤1.0 EU/μg galectin-3 |
| Product Type | Human Galectin-3, Recombinant, E.coli |
| Form | Lyophilized |
| Purity or Quality Grade | ≥95% by SDS-PAGE |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.