Learn More
Description
- Xanthine oxidase substrate
- A product of nucleic acid breakdown
- Acts to salvage free purines for nucleic acid and nucleotide recycling
- Used in conjunction with thymidine and aminopterin for cloning and selection of hybridoma cell lines
Specifications
Specifications
| CAS | 68-94-0 |
| Color | White |
| Content And Storage | Store at +15°C to +30°C |
| Form | Solid |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 136.1 |
| Purity | ≥99% (TLC) |
| Synonym | 6-Hydroxypurine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.