Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IκB Kinase Inactive Control Peptide, Cell-Permeable

Catalog No. 4014781MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-147-81MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-147-81MG Supplier MilliporeSigma™ Supplier No. 4014781MG
Add to Cart
Add to Cart

An inactive control for IκB Kinase Inhibitor Peptide.

  • Packaged Under Inert Gas
  • Peptide Sequence: Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Asp-Arg-His-Asp-Ala-Gly-Leu-Asp-Ala-Met-Lys-Asp-Glu-NH2
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white to White
Formulation Supplied as a trifluoroacetate salt.
Description 97% by HPLC
Target Proteins An inactive control for IκB Kinase Inhibitor Peptide
Molecular Formula C138H229N37O43S
Formula Weight 3126.6g/mol
Solubility H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.